missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ BRWD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550897
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The antibody detects endogenous levels of total WDR9.3 / BRWD1 protein.
WDR9.3 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 2 bromodomains and multiple WD repeats, and the function of this protein is not known. This gene is located within the Down syndrome region 2 on chromosome 21. Alternative splicing of this gene generates 3 transcript variants diverging at the 3' ends.
Specifikationer
| BRWD1 | |
| Polyclonal | |
| Unconjugated | |
| Brwd1 | |
| 5330419I02Rik; bromodomain and WD repeat domain containing 1; bromodomain and WD repeat-containing protein 1; BRWD1; C21orf107; D530019K20Rik; DCAF19; FLJ43918; G1-403-16; N143; repro5; transcriptional unit N143; WD repeat domain 9; WD repeat protein WDR9-form2; WD repeat-containing protein 9; WDR9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54014 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NSI6 | |
| Brwd1 | |
| Synthetic peptide corresponding to residues near the N terminal of human bromodomain and WD repeat domain containing 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering