missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRWD1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00054014-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MAEPSSARRPVPLIESELYFLIARYLSAGPCRRAAQVLVQELEQYQLLPKRLDWEGNEHNRSYEELVLSNKHVAPDHLLQICQRIGPMLDKEIPPSISRVTSLLGAGRQSLLRTAKGTLISpecifications
| BRWD1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_001007246.1 | |
| BRWD1 | |
| BRWD1 (NP_001007247.1, 1 a.a. ∼ 120 a.a) full-length human protein. | |
| RUO | |
| 54014 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| bromodomain and WD repeat domain containing 1 | |
| C21orf107/FLJ43918/N143/WDR9 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction