missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRWD1-IT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | BRWD1-IT2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
BRWD1-IT2 Polyclonal specifically detects BRWD1-IT2 in Human samples. It is validated for Western Blot.Specifications
| BRWD1-IT2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 257357 | |
| Synthetic peptide directed towards the C terminal of human C21orf87. Peptide sequence LEARGLPCGARRTGPRRPVREMTLPSDPERATLPNPRLGAPAVPRRGPRS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BRWD1 intronic transcript 2 (non-protein coding), C21orf87, NCRNA00257 | |
| BRWD1-IT2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title