missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRUNOL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13557-1-AP-20UL
This item is not returnable.
View return policy
Description
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternatively spliced transcript variants have been identified in this gene.Specifications
| BRUNOL5 | |
| Polyclonal | |
| Unconjugated | |
| CELF5 | |
| Bruno like protein 5, BRUNOL 5, BRUNOL5, CELF 5, CELF5, RNA binding protein BRUNOL 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 314647, 319586, 60680 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N6W0 | |
| Celf5 | |
| BRUNOL5 Fusion Protein Ag4416 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction