missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BRUNOL5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5116844

Product Code. 17249343

  • £463.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: U-251MG, U-87MG, Mouse testis Immunogen sequence: MARLTESEAR RQQQQLLQPR PSPVGSSGPE PPGGQPDGMK DLDAI.

Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternatively spliced transcript variants have been identified in this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BRUNOL5
Polyclonal
Unconjugated
CELF5
4930565A21Rik; BRUNOL5; BRUNOL-5; bruno-like 5; bruno-like 5 RNA binding protein; bruno-like 5, RNA binding protein; Bruno-like protein 5; CELF5; CELF-5; CUG-BP and ETR-3 like factor 5; CUG-BP- and ETR-3-like factor 5; CUGBP Elav-like family member 5; CUGBP, Elav-like family member 5; RGD1565016; RNA-binding protein BRUNOL-5
Rabbit
Affinity chromatography
RUO
319586, 60680
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
2.63 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8N6W0
CELF5
Recombinant fusion protein containing a sequence corresponding to amino acids 1-45 of human CELF5 (NP_0011661441).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.