Learn More
Invitrogen™ BRUNOL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113713
Description
Immunogen sequence: GVVPFPGGHP ALETVYANGL VPYPAQSPTV AETLHPAFSG VQQYTAMYPT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%; Zebrafish: 85%.
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternatively spliced transcript variants have been identified in this gene.
Specifications
| BRUNOL5 | |
| Polyclonal | |
| Unconjugated | |
| CELF5 | |
| 4930565A21Rik; BRUNOL5; BRUNOL-5; bruno-like 5; bruno-like 5 RNA binding protein; bruno-like 5, RNA binding protein; Bruno-like protein 5; CELF5; CELF-5; CUG-BP and ETR-3 like factor 5; CUG-BP- and ETR-3-like factor 5; CUGBP Elav-like family member 5; CUGBP, Elav-like family member 5; RGD1565016; RNA-binding protein BRUNOL-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 60680 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N6W0 | |
| CELF5 | |
| Synthetic peptide directed towards the middle region of human BRUNOL5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.