missing translation for 'onlineSavingsMsg'
Learn More

Brucine sulfate hydrate, 98+%, Thermo Scientific™

Product Code. 10256940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10256940 25 g Glass bottle 25g
10239493 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10256940 Supplier Thermo Scientific Acros Supplier No. 218170250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 652154-10-4
Molecular Formula C46H54N4O12S
Molecular Weight (g/mol) 887.01
MDL Number MFCD00013472
InChI Key HCMSIGALSOEZRW-MDQLRNFDSA-N
Synonym brucine sulfate salt
PubChem CID 118797651
IUPAC Name 10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;hydrate
SMILES OS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1

Specifications

Melting Point 180.0°C
Color White to Beige
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Sensitivity (to nitrate) Passes Test
Specific Rotation − 24.00
Formula Weight 443.5
Percent Purity 98+%
Insoluble Matter (in water) Passes test
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material Brucine sulfate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Brucine sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.