missing translation for 'onlineSavingsMsg'
Learn More

Brucine sulfate heptahydrate, ACS

Product Code. 15481447
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15481447 Supplier Thermo Scientific Alfa Aesar Supplier No. 036249.14
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60583-39-3 | C46H68N4O19S | 1013.12 g/mol

As Resolving agent. Used in the Determination of nitrate. Used in Pharmaceutical and food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60583-39-3
Melting Point 180°C (decomposition)
Molecular Formula C46H68N4O19S
MDL Number MFCD00150159
Quantity 25 g
UN Number UN2811
Merck Index 14,1455
Synonym brucinesulfateheptahydrate fornitrateanalysis
Solubility Information Soluble in hot water.
InChI Key PPJIVFYMRNHHTJ-UMRIXEKQSA-N
SMILES O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
IUPAC Name (4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate
Molecular Weight (g/mol) 1013.12
PubChem CID 126969999
Formula Weight 1013.15 (887.03 Anhydrous)
Grade ACS
Physical Form Powder
Chemical Name or Material Brucine sulfate heptahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Brucine sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Brucine sulfate heptahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.