missing translation for 'onlineSavingsMsg'
Learn More

Brucine dihydrate, 98%

Product Code. 11461980
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461980 10 g 10g
11471980 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461980 Supplier Thermo Scientific Alfa Aesar Supplier No. A16251.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5892-11-5 | C6H3FeK3O12 | 440.22 g/mol

Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

Solubility
Soluble in methanol. Partially soluble Miscible with water.

Notes
Incompatible with strong oxidizing agents, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5892-11-5
MDL Number MFCD00149384

Specifications

Melting Point 176°C to 178°C
Molecular Formula C6H3FeK3O12
Quantity 10 g
UN Number UN1570
Solubility Information Soluble in methanol. Partially soluble Miscible with water.
InChI Key YXTWCYYDGRBUDE-UHFFFAOYSA-K
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
IUPAC Name tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
Molecular Weight (g/mol) 440.22
PubChem CID 131675837
Formula Weight 430.50 (394.47 Anhydrous)
Percent Purity 98%
Chemical Name or Material Brucine dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.