Learn More
Brucella abortus Monoclonal Antibody (I86D), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA17391
Product Code. 11500532
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The MA1-7391 antibody reacts with Brucella abortus LPS probe in bacterial and infected tissue samples. MA1-7391 has been successfully used in ELISA and WB applications. See related products for paired capture antibody. The MA1-7391 antibody was raised against Francisella Lipopolysaccharide.
Brucella abortus is the causative agent of brucellosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Brucella abortus
Monoclonal
0.1 mg/mL
B. abortus; B. abortus Lipopolysaccharide; Brucella abortus lipopolysaccharide
Lipopolysaccharide.
100 μg
Primary
4° C
Liquid
ELISA, Western Blot
I86D
Unconjugated
Mouse
Protein A
RUO
Bacteria
Antibody
IgG1