missing translation for 'onlineSavingsMsg'
Learn More

Bruceine A, MedChemExpress

Product Code. 16481528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481528 5 mg 5mg
16439128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481528 Supplier Medchem Express Supplier No. HYN0841
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bruceine A(NSC310616; Dihydrobrusatol) is a natural quassinoid compound extracted from the dried fruits of Brucea javanica (L.

Bruceine A(NSC310616

Specifications

Chemical Name or Material Bruceine A
CAS 25514-31-2
Color White
Purity Grade Notes Research
Molecular Formula C26H34O11
Quantity 5 mg
Synonym Dihydrobrusatol NSC310616
Solubility Information DMSO : 12 mg/mL (22.96 mM; Need ultrasonic and warming)
SMILES O=C([C@@]1([C@H]([C@H](O)[C@]2([H])[C@]3(C4)C)O)[C@@]([C@H](C5=O)OC(CC(C)C)=O)([H])[C@@]2([C@@](O5)([H])C[C@@]3([H])C(C)=C(O)C4=O)CO1)OC
Molecular Weight (g/mol) 522.54
Formula Weight 522.54
Percent Purity 96.61%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.