missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRUCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567360
This item is not returnable.
View return policy
Description
Immunogen sequence: GSTDNESCTNS ELNSPLVRRT LPVLLLYSIK ESDEKAGKIF SQMNNIMSKS LHDDGFTVPQ IIEMELDSQE QLLLQDPPVT YIQQFADAA Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and can be prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family that binds to and inhibits cell death proteases. BRUCE, also known as BIRC6, is an IAP family member protein with a BIR (baculoviral inhibition of apoptosis protein repeat) domain and a UBCc (ubiquitin-conjugating enzyme E2, catalytic) domain. BRUCE regulates p53 and the mitochondrial pathway of apoptosis by facilitating the degradation of apoptotic proteins such as Caspase-9 and SMAC by ubiquitination.
Specifications
| BRUCE | |
| Polyclonal | |
| Unconjugated | |
| BIRC6 | |
| A430032G04Rik; A430040A19Rik; AA501170; APOLLON; baculoviral IAP repeat containing 6; baculoviral IAP repeat-containing 6; baculoviral IAP repeat-containing protein 6; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; accccbaculoviral IAP repeat-containing 6; BIR repeat-containing ubiquitin-conjugating enzyme; Birc6; BRUCE; D630005A10Rik; KIAA1289; mKIAA1289; RING-type E3 ubiquitin transferase BIRC6; Ubiquitin-conjugating BIR domain enzyme apollon; ubiquitin-conjugating BIR-domain enzyme apollon | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NR09 | |
| BIRC6 | |
| Recombinant Human BRUCE. Recombinant protein control fragment (Product #RP-108015). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction