missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRSK1 Polyclonal Antibody

BRSK1 Polyclonal Antibody
Brand: Invitrogen PA5101216
This item is not returnable.
View return policy
Description
BRSK1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| BRSK1 | |
| Polyclonal | |
| Unconjugated | |
| Brsk1 | |
| BR serine/threonine kinase 1; BR serine/threonine-protein kinase 1; Brain-selective kinase 1; brain-specific serine/threonine-protein kinase 1; Brsk1; Gm1100; hSAD1; KIAA1811; protein kinase SAD1A; RGD1563268; SAD1; SAD1 homolog; SAD1 kinase; SAD1A; SADB; SAD-B; SadB kinase short isoform; serine/threonine kinase SADB; serine/threonine kinase SAD-B; serine/threonine-protein kinase BRSK1; Serine/threonine-protein kinase SAD-B; synapses of Amphids Defective homolog 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 381979, 84446 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5RJI5, Q8TDC3 | |
| Brsk1 | |
| A synthesized peptide derived from human BRSK1(Accession Q8TDC3), corresponding to amino acid residues Y352-V402. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction