missing translation for 'onlineSavingsMsg'
Learn More

BRSK1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18431753 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431753 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431753 Supplier Novus Biologicals Supplier No. NBP176778AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRSK1 Polyclonal antibody specifically detects BRSK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRSK1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias BR serine/threonine kinase 1, EC 2.7.11, EC 2.7.11.1, FLJ43009, KIAA1811BR serine/threonine-protein kinase 1, protein kinase SAD1A, SAD1, SAD1 kinase, Serine/threonine-protein kinase SAD-B
Host Species Rabbit
Immunogen Antibody was raised against a 28 amino acid synthetic peptide from near the carboxy terminus of human BRSK1. The immunogen is located within amino acids 720 - 770 of BRSK1. Amino Acid Squence: DEKNGAQTRPAGAPPRSLQPPPGRPDPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84446
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.