Learn More
BRPF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551909
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SLAGDEATHH TEDAAEEERL VLLENQKHLP VEEQLKLLLE RLDEVNASKQ SVGRSRRAKM IKKEMTALRR KLAHQRETGR DGPERHGPSS RGSLTPHPAA CDKDGQTD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.Specifications
BRPF1 | |
Polyclonal | |
Unconjugated | |
BRPF1 | |
4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P55201 | |
BRPF1 | |
Recombinant protein corresponding to Human BRPF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only