missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRP44, Mouse anti-Human, Clone: 2D8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00025874-M15.100ug
This item is not returnable.
View return policy
Description
Sequence: MSAAGARGLRATYHRLLDKVELMLPEKLRPLYNHPAGPRTVFFWAPIMKW GLVCAGLADMARPAEKLSTAQSAVLMATGFIWSRYSLVIIPKNWSLFAVN FFVGAAGASQLFRIWRYNQELKAKAHK&asterisk;Specifications
| BRP44 | |
| Monoclonal | |
| Unconjugated | |
| brain protein 44 | |
| DKFZp564B167/MGC125752/MGC125753 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2D8 | |
| In 1x PBS, pH 7.4 | |
| NM_015415 | |
| BRP44 | |
| BRP44 (NP_056230, 1 a.a. ∼ 127 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 25874 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction