missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRP44 Antibody (JCM-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80086
This item is not returnable.
View return policy
Description
BRP44 Monoclonal specifically detects BRP44 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| BRP44 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunoprecipitation | |
| brain protein 44, DKFZP564B167, MGC125752, MGC125753 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 25874 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| JCM-1 | |
| Unconjugated | |
| PBS containing 25% glycerol | |
| MPC2 | |
| BRP44 peptide coupled to KLH. | |
| 100 μg | |
| Primary | |
| Recognizes endogenous human and mouse BRP44. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction