missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Broxyquinoline, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Broxyquinoline
Specifications
Specifications
| Chemical Name or Material | Broxyquinoline |
| CAS | 521-74-4 |
| Analyte or Component Names | Broxyquinoline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H5 Br2 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5,7-Dibromo-8-hydroxyquinoline, 5,7-Dibromo-8-quinolinol, 5,7-Dibromooxine, Brodiar, Broxyquinolin, Broxyquinoline, Colepur, Colipar, Dibromoksin, Dibromoquin, Dibromoxin, Dibromoxine, Dibromoxyquinoline, Fenilor, Intensopan, NSC 1810, NSC 74937, Paramiba, Paramibe, 8-Quinolinol, 5,7-dibromo- |
| InChI Formula | InChI=1S/C9H5Br2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H |
| SMILES | Oc1c(Br)cc(Br)c2cccnc12 |
| IUPAC Name | 5,7-dibromoquinolin-8-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction