missing translation for 'onlineSavingsMsg'
Learn More

Broxyquinoline, MedChemExpress

Product Code. 16458686
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16458686

Brand: Medchem Express HYB1212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Broxyquinoline (Dibromohydroxyquinoline) is a potent severe fever with thrombocytopenia syndrome virus (SFTSV) inhibitor with an IC50 of 5.8 μM. Broxyquinoline is an antiprotozoal agent.

Broxyquinoline (Dibromohydroxyquinoline) is a potent severe fever with thrombocytopenia syndrome virus (SFTSV) inhibitor with an IC50 of 5.8 μM. Broxyquinoline is an antiprotozoal agent.

Specifications

Chemical Name or Material Broxyquinoline
Physical Form Solid
CAS 521-74-4
Color Light Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H5Br2NO
For Use With (Application) COVID-19-immunoregulation
Synonym Dibromohydroxyquinoline 5,7-Dibromo-8-hydroxyquinoline
Solubility Information DMSO : 31.25 mg/mL (103.15 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.21 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=C(Br)C=C1Br
Molecular Weight (g/mol) 302.95
Formula Weight 302.95
Percent Purity 99.42%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.