Learn More
Hirschmann™ Amber Glass Stoppers
Product Code. 30090397
Change view
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
30090397 NS 10/19 Pack of 10
30090419 NS 7/16 Pack of 10
30090422 NS 12/21 Pack of 10
30090413 NS 14/23 Pack of 10
10097602 NS 29/32 Each
10252023 NS 24/29 Each
30090417 NS 19/26 Pack of 10
10417363 NS 34/35 Each
10057652 NS 45/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 30090397 Supplier Hirschmann™ Supplier No. 8902360
Only null left
Add to basket
Add to basket
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
  • Acc. to DIN 12 252
Specifications
For Use With (Equipment) For tight sealing of standard ground vessels
Quantity 1/Ea.
Standard Taper Joint NS 10/19
Material Glass
Product Type Hexagonal Head Stopper