missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brorin/VWC2 Rabbit anti-Human, Clone: 44, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Brorin/VWC2 |
|---|---|
| Clone | 44 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228346
|
Novus Biologicals
NBP2-90045-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18296506
|
Novus Biologicals
NBP2-90045-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Brorin/VWC2 Monoclonal specifically detects Brorin/VWC2 in Human samples. It is validated for ELISA.Specifications
| Brorin/VWC2 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 375567 | |
| IgG | |
| Protein A purified |
| 44 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2 | |
| Recombinant Human Brorin/VWC2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title