missing translation for 'onlineSavingsMsg'
Learn More

Brorin/VWC2 Antibody (08), Novus Biologicals™

Product Code. 18662698 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18662698 100 μg 100µL
18654998 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18662698 Supplier Novus Biologicals Supplier No. NBP305797100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Brorin/VWC2 Monoclonal antibody specifically detects Brorin/VWC2 in Human samples. It is validated for ELISA, Sandwich ELISA Capture
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brorin/VWC2
Applications ELISA, Sandwich ELISA Capture
Classification Monoclonal
Clone 08
Conjugate Unconjugated
Formulation 0.2 um filtered solution in PBS
Gene Alias Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Brorin/VWC2 (Accession#: NP_940972.2; Met1-Met325).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375567
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.