missing translation for 'onlineSavingsMsg'
Learn More

Bronze powder, -100 mesh, 99.0% (metals basis)

Product Code. 11399249
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399249 1 kg 1kg
11389249 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399249 Supplier Thermo Scientific Alfa Aesar Supplier No. 088359.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bronze
Appearance Bronze colored powder
Quantity 1 kg
Mesh Size -100 Mesh
Odor Odorless
Assay Percent Range 99% (Metals basis)
Form Powder
MDL Number MFCD00198187
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.