missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bronopol, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Bronopol |
| CAS | 52-51-7 |
| Molecular Formula | C3 H6 Br N O4 |
| Quantity | 25 g |
| Synonym | 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropan-1,3-diol, 2-Bromo-2-nitropropane-1,3-diol, 2-Nitro-2-bromo-1,3-propanediol, Acticide L 30, BE 6, BE 6 (bactericide), BNPD, BNPK, Bactrinashak, Bactrinol 100, Bactronol, Bioban, Bioban BP Plus, Bronidiol, Bronocot, Bronopol, Bronotak, Busan 1144, Canguard 409, Myacide AS, Myacide AS Plus, Myacide BT, Myacide Pharma BP, N 25, N 25 (antimicrobial), NSC 141021, Nalco 92RU093, Onyxide 500, Preventol P 100, Protectol BN 98, Protectol BN 99, Pyceze, Topcide 2520, Ultra-Fresh SAB |
| InChI Formula | InChI=1S/C3H6BrNO4/c4-3(1-6,2-7)5(8)9/h6-7H,1-2H2 |
| SMILES | OCC(Br)(CO)[N+](=O)[O-] |
| IUPAC Name | 2-bromo-2-nitropropane-1,3-diol |
| Molecular Weight (g/mol) | 199.99 |
| Formula Weight | 198.948 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction