missing translation for 'onlineSavingsMsg'
Learn More

Bronopol, MedChemExpress

Product Code. 16498686
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498686 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498686 Supplier Medchem Express Supplier No. HYB1217

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Bronopol is an antimicrobial, with low mammalian toxicity (at in-use levels) and high activity against bacteria (especially the troublesome Gram-negative species).

Bronopol is an antimicrobial, with low mammalian toxicity (at in-use levels) and high activity against bacteria (especially the troublesome Gram-negative species).

Specifications

Chemical Name or Material Bronopol
Physical Form Solid
CAS 52-51-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H6BrNO4
For Use With (Application) COVID-19-immunoregulation
Synonym BNPD BNPK
Solubility Information DMSO : 100 mg/mL (500.02 mM; Need ultrasonic) ∣H2O : 100 mg/mL (500.02 mM; Need ultrasonic)
SMILES OCC([N+]([O-])=O)(Br)CO
Molecular Weight (g/mol) 199.99
Formula Weight 199.99
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.