missing translation for 'onlineSavingsMsg'
Learn More

Brompheniramine maleate, MedChemExpress

Product Code. 16438336
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16438336

Brand: Medchem Express HYB0480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Brompheniramine ((±)-Brompheniramine) maleate is a potent and orally active antihistamine of the alkylamine class.

Brompheniramine maleate is a selective histamine H1 receptor antagonist with a Kd of 6.06 nM. Brompheniramine maleate can block the hERG channels, calcium channels, and sodium channels with IC50s of 0.90 μM, 16.12 μM and 21.26 μM, respectively. Brompheniramine maleate has anticholinergic, antidepressant and anesthetic properties and can be used for allergic rhinitis research.

Specifications

Chemical Name or Material Brompheniramine maleate
CAS 980-71-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H23BrN2O4
Quantity 100 mg
Synonym (±)-Brompheniramine maleate
Solubility Information DMSO : 100 mg/mL (229.72 mM; Need ultrasonic) ∣H2O : 100 mg/mL (229.72 mM; Need ultrasonic)
SMILES O=C(O)/C=C\C(O)=O.CN(C)CCC(C1=CC=CC=N1)C2=CC=C(Br)C=C2
Molecular Weight (g/mol) 435.31
Formula Weight 435.31
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.