missing translation for 'onlineSavingsMsg'
Learn More

Bromotriphenylmethane, 98%

Product Code. 11394569
Change view
Click to view available options
Quantity:
10 g
50 g
500 g
Unit Size:
10g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394569 10 g 10g
11304579 50 g 50g
11415293 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394569 Supplier Thermo Scientific Alfa Aesar Supplier No. A12864.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 596-43-0 | C19H15Br | 323.233 g/mol

It is considered as a more reactive reagent than Chlorotriphenyl­methane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is considered as a more reactive reagent than Chlorotriphenyl­methane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.

Solubility
Solubility in hot acetic acid is almost transparent.

Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-43-0
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.233
MDL Number MFCD00000120
InChI Key NZHXEWZGTQSYJM-UHFFFAOYSA-N
Synonym bromotriphenylmethane, triphenylmethyl bromide, trityl bromide, triphenylbromomethane, bromodiphenylmethyl benzene, methane, bromotriphenyl, benzene, 1,1',1-bromomethylidyne tris, triphenylmethylbromide, bromomethanetriyl tribenzene, 2-bromo-1,1,1-triphenylethane
PubChem CID 11692
IUPAC Name [bromo(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br

Specifications

Melting Point 152°C to 156°C
Boiling Point 230°C (15 mmHg)
Quantity 10 g
UN Number UN1759
Beilstein 1878494
Formula Weight 323.24
Percent Purity 98%
Chemical Name or Material Bromotriphenylmethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.