missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromotrichloromethane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
Specifications
Melting Point | -6°C |
Density | 2.015 |
Boiling Point | 104°C to 105°C |
Flash Point | None |
Odor | Chloroform-like |
Linear Formula | BrCCl3 |
Refractive Index | 1.506 |
Quantity | 500 g |
UN Number | UN2810 |
Beilstein | 1732543 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'