missing translation for 'onlineSavingsMsg'
Learn More

Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 97%

Product Code. 11344316
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344316 1 g 1g
11358666 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344316 Supplier Thermo Scientific Alfa Aesar Supplier No. L19384.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132705-51-2 | C12H24BrF6N3P2 | 466.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132705-51-2
Molecular Formula C12H24BrF6N3P2
Molecular Weight (g/mol) 466.187
MDL Number MFCD00077412
InChI Key CYKRMWNZYOIJCH-UHFFFAOYSA-N
Synonym pybrop, bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, bromotripyrrolidinophosphonium hexafluorophosphate, bromotri pyrrolidin-1-yl phosphonium hexafluorophosphate v, pybrop r, bromotri 1-pyrrolidinyl phosphonium hexafluorophosphate, bromotri pyrrolidino phosphonium hexafluorophosphate, bromo-tris-pyrrolidinophosphoniumhexafluorophosphate, bromo-tris-pyrrolidino phosphoniumhexafluorophosphate, bromo-tris-pyrrolidino-phosphoniumhexafluorophosphate
PubChem CID 2733179
IUPAC Name bromo(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Br.F[P-](F)(F)(F)(F)F

Specifications

Quantity 1 g
Beilstein 7845702
Sensitivity Moisture sensitive
Formula Weight 466.19
Percent Purity 97%
Chemical Name or Material Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.