missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, sodium salt, pure, water soluble, indicator

Product Code. 10711555 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711555 100 g Glass bottle 100g
10487360 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711555 Supplier Thermo Scientific Chemicals Supplier No. 151371000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
Synonym Bromthymol Blue, sodium salt, 3', 3''-Dibromothymolsulfonephthalein, sodium salt, BTB
PubChem CID 102183223
IUPAC Name sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Merck Index 15, 1455
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 646.35
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Visual Transition Interval from pH 6.0 (yellow) to pH 7.6 (blue)
Color Brown/Orange to Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Bromothymol Blue, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.