missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, free acid, For ACS analysis, MP Biomedicals™

Product Code. 11457801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457801 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457801 Supplier MP Biomedicals Supplier No. 0215052425
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Synonym 3', 3''-Dibromothymolsulfone-phthalein
Color Lilac
Quantity 25 g
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Physical Form Powder
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Molecular Weight (g/mol) 624.384
PubChem CID 6450
ChEBI CHEBI:86155
Formula Weight 624.4
Chemical Name or Material Bromothymol Blue, free acid, For ACS analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.