missing translation for 'onlineSavingsMsg'
Learn More

β-Bromostyrene, 97%, mixture of cis/trans isomers, Thermo Scientific Chemicals

Product Code. 10675032
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-64-0
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000185
InChI Key YMOONIIMQBGTDU-VOTSOKGWSA-N
Synonym 2-bromovinyl benzene, beta-bromostyrene, styryl bromide, 2-bromoethenyl benzene, bromostyrolene, 1-bromo-2-phenylethene, bromostyrol, bromstyrole, hyacinth base, 1-bromo-2-phenylethylene
PubChem CID 5314126
IUPAC Name [(E)-2-bromoethenyl]benzene
SMILES C1=CC=C(C=C1)C=CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 7.0°C
Color Yellow to Brown
Density 1.4200g/mL
Boiling Point 110.0°C to 112.0°C (20.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (sum of isomers) (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHBr
Refractive Index 1.6060 to 1.6090
Quantity 5 mL
Beilstein 05, 477
Specific Gravity 1.42
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 183.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material β-Bromostyrene, Mixture of cis and trans
Show More Show Less
compliance-icons
Product Identifier
  • ß-Bromostyrene, mixture of cis/trans isomers
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'