missing translation for 'onlineSavingsMsg'
Learn More

Bromosporine, MedChemExpress

Product Code. 16489387
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489387 5 mg 5mg
16446848 10 mg 10mg
16442917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489387 Supplier Medchem Express Supplier No. HY15815
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bromosporine is a broad spectrum inhibitor for bromodomains with IC50 of 0.41 (mu)M, 0.29 (mu)M, 0.122 (mu)M and 0.017 (mu)M for BRD2, BRD4, BRD9 and CECR2, respectively.

Bromosporine is a broad spectrum inhibitor for bromodomains with IC50 of 0.41 (mu)M, 0.29 (mu)M, 0.122 (mu)M and 0.017 (mu)M for BRD2, BRD4, BRD9 and CECR2, respectively.

Specifications

Chemical Name or Material Bromosporine
CAS 1619994-69-2
Color White
Purity Grade Notes Research
Molecular Formula C17H20N6O4S
Quantity 5 mg
Solubility Information DMSO : ≥ 51.7 mg/mL (127.83 mM)
SMILES O=C(OCC)NC1=CC(C2=CC=C(C)C(NS(=O)(C)=O)=C2)=NN3C1=NN=C3C
Molecular Weight (g/mol) 404.44
Formula Weight 404.44
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.