missing translation for 'onlineSavingsMsg'
Learn More

Bromopyruvic Acid, TRC

Product Code. 30133351
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133351 5 g 5g
30133941 25 g 25g
17992585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133351 Supplier Toronto Research Chemicals Supplier No. TRCB6869305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-59-3
Molecular Formula C3H3BrO3
Molecular Weight (g/mol) 166.96
Synonym 3-Bromo-2-oxopropanoic Acid, 3-Bromopyruvate, 3-Bromopyruvic Acid, NSC 11731, NSC 62343, β-Bromopyruvic Acid;
IUPAC Name 3-bromo-2-oxopropanoic acid
SMILES C(C(=O)C(=O)O)Br

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H3BrO3/c4-1-2(5)3(6)7/h1H2,(H,6,7)
Formula Weight 165.9266
Chemical Name or Material Bromopyruvic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.