missing translation for 'onlineSavingsMsg'
Learn More

Bromopyruvic acid, 97%

Product Code. 11370184
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370184 50 g 50g
11350184 2 g 2g
11360184 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11370184 Supplier Thermo Scientific Alfa Aesar Supplier No. L00720.18
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1113-59-3 | C3H3BrO3 | 166.958 g/mol

Bromopyruvic acid is used as an affinity label for cysteine residues and a potential anti-cancer agent. It is involved in the synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids and substituted pyranones. It plays a vital role as a cross-linker between nucleic acids and proteins. It is used as an anticancer agent for lung tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromopyruvic acid is used as an affinity label for cysteine residues and a potential anti-cancer agent. It is involved in the synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids and substituted pyranones. It plays a vital role as a cross-linker between nucleic acids and proteins. It is used as an anticancer agent for lung tumors.

Solubility
Soluble in water, dimethyl sulfoxide, ethyl acetate and water.

Notes
Moisture and air sensitive. Store in cool place. Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-59-3
Molecular Formula C3H3BrO3
Molecular Weight (g/mol) 166.958
MDL Number MFCD00002587
InChI Key PRRZDZJYSJLDBS-UHFFFAOYSA-N
Synonym 3-bromopyruvic acid, bromopyruvic acid, bromopyruvate, 3-bromopyruvate, 3-bromo-2-oxopropionic acid, bromopyruvicacid, pyruvic acid, bromo, propanoic acid, 3-bromo-2-oxo, unii-63jmv04grk, .beta.-bromopyruvic acid
PubChem CID 70684
IUPAC Name 3-bromo-2-oxopropanoic acid
SMILES C(C(=O)C(=O)O)Br

Specifications

Melting Point 75°C to 80°C
Quantity 50 g
UN Number UN3261
Beilstein 1746786
Solubility Information Soluble in water,dimethyl sulfoxide,ethyl acetate and water.
Formula Weight 166.96
Percent Purity 97%
Chemical Name or Material Bromopyruvic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.