missing translation for 'onlineSavingsMsg'
Learn More

Bromopyramine Hydrochloride (N-[(4-Bromophenyl)methyl]-N-[2-(dimethylamino)ethyl]pyridin-2-amine Hydrochloride), LoGiCal

Product Code. 30461476
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30461476

Brand: LoGiCal LGCFOR1894.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Bromopyramine Hydrochloride (N-[(4-Bromophenyl)methyl]-N-[2-(dimethylamino)ethyl]pyridin-2-amine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromopyramine Hydrochloride
CAS 1089707-20-9
Analyte or Component Names Bromopyramine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H20 Br N3 . Cl H
Certifications/Compliance ISO 9001
Synonym 1,2-Ethanediamine, N1-[(4-bromophenyl)methyl]-N2,N2-dimethyl-N1-2-pyridinyl-, hydrochloride (1:1), Bromopyramine Hydrochloride, N-[(4-Bromophenyl)methyl]-N-[2-(dimethylamino)ethyl]pyridin-2-amine Hydrochloride
InChI Formula InChI=1 S/C16H20BrN3.ClH/c1-19(2)11-12-20(16-5-3-4-10-18-16)13-14-6-8-15(17)9-7-14;/h3-10 H,11-13H2,1-2H3;1 H
SMILES Cl.CN(C)CCN(Cc1ccc(Br)cc1)c2ccccn2
IUPAC Name N'-[(4-bromophenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
Industry Type Forensics
Molecular Weight (g/mol) 370.72
Quantity 5 mg
Formula Weight 369.0607 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.