missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Red, Pure, Indicator Grade, Fisher Chemical

Product Code. 12347133 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12347133 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12347133 Supplier Fisher Chemical Supplier No. B/4660/43
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2800-80-8
Molecular Formula C19H12Br2O5S
MDL Number 23014
Formula Weight 512.19g/mol
Packaging Glass Jar
Color Brown/Red
Quantity 1 g
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.