missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Red, pure

Product Code. 10043660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10043660 5 g 5g
10359310 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10043660 Supplier Thermo Scientific Chemicals Supplier No. 304550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2800-80-8 | C19H12Br2O5S | 512.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2800-80-8
Molecular Formula C19H12Br2O5S
Molecular Weight (g/mol) 512.168
MDL Number MFCD00023014
InChI Key OYCLSQDXZMROJK-UHFFFAOYSA-N
Synonym 5', 5''-Dibromophenolsulfonphthalein
PubChem CID 76047
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br

Specifications

Infrared Spectrum Authentic
Formula Weight 512.19
Loss on Drying 5% max. (110°C)
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 5.2 (yellow-red) to pH 7.0 (purple)
Color Brown to Brown/Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromophenol Red

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.