missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent

Product Code. 10701555
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.94
MDL Number MFCD00013793
InChI Key KGANHHDZDYZJEM-UHFFFAOYNA-M
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein
PubChem CID 3791690
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate

Specifications

Melting Point >300.0°C
Beilstein 19, I, 649
SMILES [Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
Formula Weight 691.94
Grade Pure
Packaging Glass bottle
Quantity 100 g
Chemical Name or Material Bromophenol Blue, sodium salt, Pure

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'