missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, sodium salt, pure, water soluble, indicator

Product Code. 10380904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10380904 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10380904 Supplier Thermo Scientific Chemicals Supplier No. 151350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-28-9 | C19H9Br4NaO5S | 691.942 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Melting Point >300.0°C
CAS 62625-28-9
Molecular Formula C19H9Br4NaO5S
MDL Number MFCD00013793
Beilstein 19, I, 649
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 691.942
PubChem CID 3791690
Formula Weight 691.94
Grade Pure
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Bromophenol Blue, sodium salt, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.