missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue sodium salt

Product Code. 11478716
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478716 50 g 50g
11468716 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478716 Supplier Thermo Scientific Chemicals Supplier No. A16899.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34725-61-6 | C19H9Br4NaO5S | 691.96 g/mol

Bromophenol blue sodium salt is a tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels. Keep away from strong oxidizing agents. It is used in analytical chemistry as Ph indicators. It is also used as a tracking dye for alkaline and neutral buffer systems. It is used as an acid-base indicator and a gel electrophoresis color marker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromophenol blue sodium salt is a tracking dye for protein or nucleic acid electrophoresis in polyacrylamide or agarose gels. Keep away from strong oxidizing agents. It is used in analytical chemistry as Ph indicators. It is also used as a tracking dye for alkaline and neutral buffer systems. It is used as an acid-base indicator and a gel electrophoresis color marker.

Solubility
Soluble in water and slightly soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34725-61-6
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.96
MDL Number MFCD00013793
Synonym Bromphenol Blue, water soluble

Specifications

Melting Point >300°C
Merck Index 141444
Solubility Information Soluble in water and slightly soluble in alcohol.
Formula Weight 691.96
Quantity 50 g
Chemical Name or Material Bromophenol Blue sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.