missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, pure, indicator

Product Code. 10564264 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10564264 100 g Glass bottle 100g
10551591 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10564264 Supplier Thermo Scientific Chemicals Supplier No. 151341000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bromophenol Blue, C19H10Br4O5S, CAS Number-115-39-9, 3', 3″, 5', 3',3″,5',5″-Tetrabromophenolsulfonephthalein, BPB, 25g, 19, I, 649, CHEBI:59424, Orange-Pink to Red, 669.96, Pure, UDSAIICHUKSCKT-UHFFFAOYSA-N, Authentic | CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 279.0°C
Beilstein 19, I, 649
Merck Index 15, 1454
Solubility Information Solubility in water: 0.4g/100mL
Infrared Spectrum Authentic
Formula Weight 669.96
Grade Pure
Packaging Glass bottle
Specific Extinction 1100 min. (592nm, in buffer)
Visual Transition Interval from pH 3.0 (yellow) to pH 4.6 (blue)
Color Orange/Pink to Red
Quantity 100 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue, Indicator
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.