Learn More
Bromopentamethylbenzene, 98%
Description
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Tekniske data
Tekniske data
| Melting Point | 161°C to 162°C |
| Quantity | 5 g |
| Beilstein | 1863555 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 227.15 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Bromopentamethylbenzene |
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.