missing translation for 'onlineSavingsMsg'
Learn More

Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals

Product Code. 10341094 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341094 1 g Glass bottle 1g
10165700 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341094 Supplier Thermo Scientific Acros Supplier No. 307050010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-70-2
Molecular Formula CH2BrNO2
Molecular Weight (g/mol) 139.94
MDL Number MFCD00007401
InChI Key DNPRVXJGNANVCZ-UHFFFAOYSA-N
Synonym methane, bromonitro, bromo nitro methane, nitrobromomethane, unii-eg63p9eur0, eg63p9eur0, ccris 9456, bromo-nitromethane, 1-bromonitromethane, methane,bromonitro, monobromonitromethane
PubChem CID 68423
IUPAC Name bromo(nitro)methane
SMILES C([N+](=O)[O-])Br

Specifications

Color Yellow
Density 2.0700g/mL
Boiling Point 146°C to 148°C (750.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2NO2
Refractive Index 1.4850 to 1.495
Quantity 1 g
Specific Gravity 2.07
Formula Weight 139.94
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Bromonitromethane, tech., 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.