missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)triphenylphosphonium bromide, 98%

Product Code. 10782594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10782594 25 g Glass bottle 25g
10687641 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10782594 Supplier Thermo Scientific Acros Supplier No. 321480250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1034-49-7 | C19H17Br2P | 436.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1034-49-7
Molecular Formula C19H17Br2P
Molecular Weight (g/mol) 436.13
MDL Number MFCD00011864
InChI Key YFTMLUSIDVFTKU-UHFFFAOYSA-M
Synonym bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
PubChem CID 2733422
IUPAC Name bromomethyl(triphenyl)phosphanium;bromide
SMILES [Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C to 235.0°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2P(C6H5)3Br
Quantity 25 g
Formula Weight 436.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Bromomethyl)triphenylphosphonium bromide
Show More Show Less
compliance-icons
Product Identifier
  • (Bromomethyl)triphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (Bromomethyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.