missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl methyl ether, tech. 90%

Product Code. 11362287
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11362287

Brand: Thermo Scientific Alfa Aesar L09837.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13057-17-5 | C2H5BrO | 124.965 g/mol

Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.

Solubility
Sparingly soluble in water.

Notes
Moisture Sensitive. Store at 4°C. Do not store together with acids. Store away from water and strong bases. Incompatible with acids, bases, moisture, heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13057-17-5
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.965
MDL Number MFCD00000171
InChI Key JAMFGQBENKSWOF-UHFFFAOYSA-N
Synonym bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether
PubChem CID 83093
IUPAC Name bromo(methoxy)methane
SMILES COCBr

Specifications

Density 1.531
Boiling Point 86°C to 87°C
Flash Point 26°C (78°F)
Odor Pungent
Refractive Index 1.4520-1.4600
Quantity 25 g
UN Number UN1992
Beilstein 1730853
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.
Formula Weight 124.97
Percent Purity 90%
Grade Technical
Chemical Name or Material Bromomethyl methyl ether
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.