missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl methyl ether, tech. 90%

Product Code. 11352287
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352287 5 g 5g
11362287 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352287 Supplier Thermo Scientific Alfa Aesar Supplier No. L09837.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13057-17-5 | C2H5BrO | 124.965 g/mol

Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.

Solubility
Sparingly soluble in water.

Notes
Moisture Sensitive. Store at 4°C. Do not store together with acids. Store away from water and strong bases. Incompatible with acids, bases, moisture, heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13057-17-5
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.965
MDL Number MFCD00000171
InChI Key JAMFGQBENKSWOF-UHFFFAOYSA-N
Synonym bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether
PubChem CID 83093
IUPAC Name bromo(methoxy)methane
SMILES COCBr

Specifications

Density 1.531
Boiling Point 86°C to 87°C
Flash Point 26°C (78°F)
Odor Pungent
Refractive Index 1.4520-1.4600
Quantity 5 g
UN Number UN1992
Beilstein 1730853
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.
Formula Weight 124.97
Percent Purity 90%
Grade Technical
Chemical Name or Material Bromomethyl methyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.