missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl methyl ether, 90%

Product Code. 12624947 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12624947 25 g Glass bottle 25g
12614947 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12624947 Supplier Thermo Scientific Acros Supplier No. 171650250
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13057-17-5 | C2H5BrO | 124.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13057-17-5
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.97
InChI Key JAMFGQBENKSWOF-UHFFFAOYSA-N
Synonym bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether
PubChem CID 83093
IUPAC Name bromo(methoxy)methane
SMILES COCBr

Specifications

Density 1.5300g/mL
Boiling Point 87.0°C
Flash Point 26°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2OCH3
Refractive Index 1.455
Quantity 25 g
Beilstein 01,582
Fieser 10,56
Specific Gravity 1.53
Formula Weight 124.97
Percent Purity 90%
Chemical Name or Material Bromomethyl methyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromomethyl methyl ether
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.