missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclobutane, 99%

Product Code. 10223052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10223052 5 mL Glass bottle 5mL
10384642 25 mL Glass bottle 25mL
10544464 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10223052 Supplier Thermo Scientific Acros Supplier No. 367640050
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17247-58-4 | C5H9Br | 149.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17247-58-4
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00040960
InChI Key FLHFTXCMKFVKRP-UHFFFAOYSA-N
Synonym bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
PubChem CID 2734658
IUPAC Name bromomethylcyclobutane
SMILES C1CC(C1)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.3390g/mL
Boiling Point 145.0°C to 148.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C4H7CH2Br
Refractive Index 1.4790 to 1.4830
Quantity 5 mL
Beilstein 05, 22
Specific Gravity 1.339
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohols
Formula Weight 149.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (Bromomethyl)cyclobutane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (Bromomethyl)cyclobutane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.