missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl acetate, 95%, Thermo Scientific Chemicals

Catalog Number 10205054
37.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
590-97-6
C3H5BrO2
152.98
MFCD00000170
NHYXMAKLBXBVEO-UHFFFAOYSA-N
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
68536
bromomethyl acetate
CC(=O)OCBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
590-97-6
C3H5BrO2
152.98
MFCD00000170
NHYXMAKLBXBVEO-UHFFFAOYSA-N
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
68536
bromomethyl acetate
CC(=O)OCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Beige
Density 1.5600g/mL
Boiling Point 130.0°C to 133.0°C (750.0 mmHg)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2Br
Refractive Index 1.4470 to 1.4540 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.56
Formula Weight 152.97
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Bromomethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bromomethyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'